Blood and tissue DNA kit, CE IVD, Mag-Bind®
Lieferant: OMEGA BIO-TEK
Mag-Bind® Blood and Tissue DNA Kit CE IVD offers a versatile method for the isolation of high-quality DNA from a wide variety of samples including fresh or frozen animal cultured cells and tissues, up to 250 µl whole blood, buccal swabs, up to 500 µl saliva, and dried blood spots.
Mag-Bind® Blood and Tissue DNA Kit CE IVD offers a versatile method for the isolation of high-quality DNA from a wide variety of samples including fresh or frozen animal cultured cells and tissues, up to 250 µl whole blood, buccal swabs, up to 500 µl saliva, and dried blood spots. Mag-Bind® Particles HDQ provide a quick magnetic response time reducing overall processing time. This system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with the time-proven efficiency of Omega Bio-Tek’s buffer chemistries to provide a fast and convenient method to isolate DNA from a variety of samples. The purification procedure provides high-quality DNA that is suitable for direct use in most downstream applications, such as amplification, next generation sequencing and enzymatic reactions.
Standards: CE-IVD IVDR