Malachitgrün-Oxalat, technische Qualität, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ200
The release of phosphate or pyrophosphate in an enzymatic reaction can be detected by malachite green and is measured spectrometrically at 660 nm.
- Malachite Green oxalate salt has been used in biliary phospholipid analysis
- Malachite Green oxalate salt has been used in the preparation of malachite green-ammonium molybdate reagent, as a stop solution for starch synthase activity assay
- To terminate the phosphatase and tensin homologue deleted on chromosome 10 (PTEN) inhibition assay using phosphatidylinositol (3,4,5) triphosphate (PI(3,4,5)P3) as substrate
- To determine trypanothione synthetase activity to measure inorganic phosphate release from adenosine triphosphate (ATP) during catalysis
Malachite green is used in the treatment of fungal and parasitic infections in fish.
Formel:
C₅₂H₅₄N₄O₁₂ Schmelzpunkt: ∼159 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00011766 CAS-Nummer: 2437-29-8 EINECS: 219-441-7 UN: 2811 ADR: 6.1,III Merck Index: 13,05722 |
|
Spezifikation Testergebnisse
Appearance (Colour) |
Metallic Green Conforms |
Appearance (Form) | Powder or Solid or Crystals or Crystalline Chunk(s) or Chunk(s) |
Solubility (Colour) 1 mg/ml, H₂O |
Dark Blue to Dark Green |
Carbon | 60.5 - 68.6% |
Nitrogen | 5.3 - 6.6% |
UV-Vis Spectrum in H₂O |
616 - 620 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...