Methylblau, Pulver und Kristalle, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Baumwollblau, Acid Blue 93, Anilinblau WS, Wasserblau 6B, Aniline blue water soluble, Anilinblau wasserlöslich, Aniline blue WS, Dinatrium-[[4-[bis[4-[(sulfonatophenyl)amino]phenyl]methylen]cyclohexa-2,5-dien-1-yliden]amino]benzolsulfonat
M-Clarity™ Qualitätsstufe = MQ200
Methyl Blue is a triaminotriphenylmethane dye, widely used in paper, textile and leather production.
- Methyl Blue has been used
- To stain leaf disks for callose visualization within the cells
- To incubate cells for 4′,6-diamidino-2-phenylindole (DAPI)/septa staining
- As a marker for β-(1-3)-d-glucans or acidic phloroglucinol for the detection of callose and lignin
Methyl Blue is considered as a xenobiotic compound and has antiseptic property. It is useful in polychrome staining. Methyl blue is mostly used for staining samples in histology and microbiology. It is also useful as a model to reveal the role of catalyst in light induced degradation of dyes.
Formel:
C₃₇H₂₇N₃Na₂O₉S₃ Molecular Weight: 799,81 g/mol Schmelzpunkt: ≥23 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00058509 CAS-Nummer: 28983-56-4 EINECS: 249-352-9 Merck Index: 13,06055 |
Spezifikation Testergebnisse
Appearance (Colour) | Dark Blue to Dark Brown |
Appearance (Form) | Powder or Crystals |
Solubility (Colour) 1 mg/ml in H2O |
Dark Blue |
E1% At Lambda Max 594 - 610 nm |
≥650 |
Wavelength | 594 - 610 nm |
Ratio P-15/P+15 Solvent: 0.005N HCl |
0.96 - 1.09 |
Recommended Retest Period | 4 years |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...