Magnesiumsulfat, anhydrous 99,5, reagent

Lieferant: Honeywell Chemicals

Synonyme: MgSO4, Magnesium sulphate anhydrous

M7506-1KG M7506-500G
M7506-1KGEA 241.54 EUR
M7506-1KG M7506-500G
Magnesiumsulfat, anhydrous 99,5, reagent
Magnesium sulphate

  • ≥95% for general laboratory use.

Formel: MgSO₄
Molecular Weight: 120.37 g/mol
Schmelzpunkt: 1.130 °C (Zers.)
Dichte: 2.7 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011110
CAS-Nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated to the dried substance) Min. 99.5 %
Loss on drying (105°C) Max. 2 %
Colour of the solution complying
Clearness of the solution complying

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR