Magnesiumsulfat, wasserfrei ≥99,5%, Reagenzqualität
Lieferant: Honeywell Chemicals
Synonyme:
MgSO4, Magnesium sulphate anhydrous
M7506-500G
M7506-500GEA
130.51
EUR
M7506-500G
Magnesiumsulfat, wasserfrei ≥99,5%, Reagenzqualität
Magnesium sulphate
- ≥95% for general laboratory use.
Formel:
MgSO₄ Molecular Weight: 120,37 g/mol Schmelzpunkt: 1,130 °C (Zers.) Dichte: 2,7 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011110 CAS-Nummer: 7487-88-9 EINECS: 231-298-2 Merck Index: 12,05731 |
Spezifikation Testergebnisse
Assay (calculated to the dried substance) | Min. 99.5 % |
Loss on drying (105°C) | Max. 2 % |
Colour of the solution | complying |
Clearness of the solution | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...