Guanidiniumchlorid solution_SC 6 M, GenAR® Biotechnology, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

Synonyme: Carbamidin Hydrochlorid, Guanidin-Chlorid, Guanidinhydrochlorid, Aminomethanamidin Hydrochlorid, Aminomethansäureamidiniumchlorid

C0563-10EA 951.28 EUR
C0563-10 0563-20 MACR0563-26
Guanidiniumchlorid solution_SC 6 M, GenAR® Biotechnology, Macron Fine Chemicals™
Guanidiniumchlorid
Formel: NH₂C(=NH)NH₂·HCl
Molecular Weight: 95.53 g/mol
Schmelzpunkt: 185
Lagertemperatur: Raumtemperatur
MDL: MFCD00013026
CAS-Nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (clear, colorless liquid) Passes Test
Cyanide (CN) Passes Test
Heavy Metals (as Pb) ≤10 ppm
Identification (by FTIR) Passes Test
ACS - Iron (Fe) ≤3 ppm
Molarity (by titrimetry) 5.8 - 6.2
pH (1:10 dilution) 4.5 - 6.5
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR