L(+)-Arginin 98,5-101,0% USP, Multikompendial, erfüllt die analytischen Spezifikationen von BP, Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

MACR4926-06EA 0 EUR
MACR4926-06 MACR4926-07 MACR4926-09
L(+)-Arginin 98,5-101,0% USP, Multikompendial, erfüllt die analytischen Spezifikationen von BP, Ph. Eur., Macron Fine Chemicals™
L(+)-Arginin
Formel: C₆H₁₄N₄O₂
Siedepunkt: 368 °C
Schmelzpunkt: 238 °C
Dichte: 1,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002635
CAS-Nummer: 74-79-3
EINECS: 200-811-1

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C6Hl4N402) (dried basis) 98.5 - 101.5%
USP - Identification Passes Test
USP - Specific Rotation (A]25 AO (+) 26.3 - 27.7°
USP - Loss on Orying at 105 °C ≤0.5%
USP - Chloride (Cl) ≤0.05%
USP - Residue on Ignition ≤0.3%
USP - Sulfate (S04) ≤0.03%
USP - Iron (Fe) ≤0.003%
USP - Related Compounds - Individual Impurities ≤0.5%
USP - Related Compounds - Total Impurities ≤2.0%
EP/BP - Assay (C6Hl4N402) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation (A]2°AO (+) 25.5 - 28.5 °
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Chloride (Cl) ≤200ppm
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Orying ≤0.5%
EP/BP - Ash (sulfated) ≤0.1%
JP - Identification Passes Test
JP - 0ptical rotation, 20 °C (+) 26.9 - 27.9°
JP - pH (1:10) 10.5 - 12.0
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021%
JP - Sulfate (S04) ≤0.028%
JP - Ammonium (NH4) ≤0.02%
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Iron (Fe) ≤10 ppm
JP - Related Substances Passes Test
JP - Loss on Orying at 105 °C ≤0.30%
JP - Residue after Ignition ≤0.1%
JP - Assay (C6Hl4N402)(dried basis) 98.5 - 101.0%
Endotoxin Concentration, IU/mg, For Information only
ChP - Specific Rotation @ 20 °C 26.9 - 27.9
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Alkalinity (pH) 10.5 - 12.0
ChP - Transmittance of solution 0.98
ChP - Chlorides ≤0.02%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - Proteins Passes Test
ChP - 0ther amino acids (TLC)(<=0.4%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤0.5%
ChP - Residue on ignition ≤0.1%
ChP - Iron ≤0.001%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Assay, dried basis 99.0 - 101.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR