L(+)-Histidin 98,5-101,5 Dried USP, Multi-Comp, meetSpec_BP_ChP_Ph.Eur_JP, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure, H, (S)-(+)-Histidin, His, (2S)-(+)-Histidin, H-His-OH, L-3-Imidazol-4-ylalanin

M.O.C. Category R

MACR4928-06EA 0 EUR
MACR4928-06 MACR4928-07
L(+)-Histidin 98,5-101,5 Dried USP, Multi-Comp, meetSpec_BP_ChP_Ph.Eur_JP, Macron Fine Chemicals™
L(+)-Histidin
Formel: C₆H₉N₃O₂
Molecular Weight: 155.16 g/mol
Schmelzpunkt: 287 °C
Dichte: 1,44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064315
CAS-Nummer: 71-00-1
Merck Index: 13,04741

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C H N 0 ) (dried basis) 98.5 - 101.5%
USP - Identification Passes Test
USP - Specific Rotation [A)25 AO (+) 12.6 - 1.4.0°
USP - pH 7.0 - 8.5
USP - Loss on Orying at 105 °C ≤0.2%
USP - Residue on Ignition ≤0..4%
USP - Chloride (Cl) ≤0.05%
USP - Sulfate (S04) ≤0.03%
USP - Iron (Fe) ≤0.003%
USP - Related Compounds - Individual Impurities ≤0.5%
USP - Related Compounds - Total Impurities ≤2.0%
EP/BP - Assay (C H N 0 ) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [A)2°AO (+) 11..4 - 12..4°
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Orying ≤0.5%
EP/BP - Ash (sulfated) ≤0.1%
JP - Assay (C6H9N302) (dried basis) 99.0 - 101.0%
JP - Ammonium (NH4) ≤0.02%
JP - Chloride (Cl) ≤0.021%
JP - Clarity and Color of Solution Passes Test
JP - Iron (Fe) ≤10 ppm
JP - Identification Passes Test
JP - Loss on Orying at 105 °C ≤0.30%
JP- 0ptical Rotation [A)2°AO (+) 11.8 - 12.8°
JP - Related Substances Passes Test
JP - pH 7.0 - 8.5
JP - Residue on Ignition ≤0.1%
JP - Sulfate (S04) ≤0.028%
ChP - Assay (dried basis) 99.0 - 101.0%
ChP -Specific Rotation @ 20 °C (+) 12.0 - 12.8°
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Acidity or alkalinity (pH) 7.0 - 8.5
ChP - Transmittance of solution ≥ 98.0%
ChP - Chlorides ≤0.02%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - 0ther amino acids (TLC)(<=0.5%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤0.2%
ChP - Residue on ignition ≤0.1%
ChP - Iron ≤0.001%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Bacterial endotoxin, EU/g ≤6.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR