L(+)-Histidin Hydrochlorid Monohydrat 98,5-101,0, Multi-Comp, meetSpec_BP_ChP_Ph.Eur_JP, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: His HCl Monohydrat, (S)-(+)-Histidin Hydrochlorid Monohydrat, (2S)-(+)-Histidin Hydrochlorid Monohydrat, (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat, L-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat, H-His-OH.HCl.H2O

MACR4938-06EA 0 EUR
MACR4938-06 MACR4938-07 MACR4938-09
L(+)-Histidin Hydrochlorid Monohydrat 98,5-101,0, Multi-Comp, meetSpec_BP_ChP_Ph.Eur_JP, Macron Fine Chemicals™
L(+)-Histidin Hydrochlorid Monohydrat
Formel: C₆H₉N₃O₂·HCl·H₂O
Molecular Weight: 209.63 g/mol
Schmelzpunkt: 259 °C
Dichte: 1,485 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151027
CAS-Nummer: 5934-29-2
Merck Index: 13,04741

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
EP/BP - Assay (C6H9N302 HCl) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification F Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 3.0 - 5.0
EP/BP - Specific Rotation (.A]2°AO (+) 9.2 - 10.6°
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying at 150 °C 7.0 - 10.0%
EP/BP - Ash (sulfated) ≤0.1%
JP - Identification Passes Test
JP - 0ptical Rotation (+) 9.2 - 10.6
JP - pH 3.5 - 4.5
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (S04) ≤0.028%
JP - Ammonium (NH4) ≤0.02%
JP - Iron (Fe) ≤10 ppm
JP - Related Substances Passes Test
JP - Water 7.2 - 10.0%
JP - Residue on Ignition ≤0.10%
JP - Assay (anhydrous basis) 99.0 - 101.0%
ChP - Specific Rotation @ 20 °C 8.5 - 10.5°
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Acidity or alkalinity (pH) 3.5 - 4.5
ChP - Transmittance of solution ≥ 98.0%
ChP - Chloride Content 16.7 - 17.1%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - 0ther amino acids (TLC)(<=0.2%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤0.2%
ChP - Residue on ignition ≤0.1%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Bacterial endotoxin, EU/g ≤6.0
ChP - Assay, dried basis 99.0 - 101.0%
ChP - Iron ≤0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR