E/E absolut, wasserfrei ≥99,8% (durch GC), AR®, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Ethyl alcohol

MACR6777-16EA 0 EUR
MACR6777-16
E/E absolut, wasserfrei ≥99,8% (durch GC), AR®, Macron Fine Chemicals™
Ethanol
Formel: C₂H₅OH
Molecular Weight: 46.07 g/mol
Siedepunkt: 78,3
Schmelzpunkt: -117
Flammpunkt: 12
Lagertemperatur: Raumtemperatur
MDL: MFCD00003568
CAS-Nummer: 64-17-5
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Bestellen

Spezifikation Testergebnisse

Assay (GC) ≥99.8%
Water (KF) ≤0.2%
Titrable Acid (meq/g) ≤0.0002
Residue after Evaporation ≤0.001%
Aldehydes and ketones (as C₃H₆O) ≤0.001%
Higher alcohols (as amyl alcohol) ≤0.015%
Methyl Alcohol ≤0.01%
Time for reducing KMnO4 solution ≥10 mins
Substances Darkened by H₂SO₄ Passes test
Carbonyl compounds (as CO) ≤0.003%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR