Natriummetabisulfit 65,0 - 67,4%, granular NF, FCC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Natriumpyrosulfit, Pyrosulfit, Natriumdisulfit, di-Natriumdisulfit, Disulfit

7776-26EA 3098.46 EUR
7776-26 MACR7776-06
Natriummetabisulfit 65,0 - 67,4%, granular NF, FCC, Macron Fine Chemicals™
Natriummetabisulfit
Formel: Na₂S₂O₅
Molecular Weight: 190.11 g/mol
Schmelzpunkt: 150
Lagertemperatur: Raumtemperatur
MDL: MFCD00167602
CAS-Nummer: 7681-57-4
EINECS: 231-673-0
Merck Index: 13,08712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product
Meets N.F. Requirements
GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Assay (SO₂) 65.0 - 67.4 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Thiosulfate (S₂O₃) ≤0.05 %
NF - Iron (Fe) ≤0.002 %
NF - Chloride (Cl) ≤0.05 %
FCC - Assay (Na₂S₂O₅) 96.5 - 100.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Iron (Fe) ≤0.002 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Selenium (Se) ≤5 mg/kg
Solubility (10 in 100) Passes Test
Appearance (white or yellowish white crystalline granules)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR