Chlortrimethylsilan Silylierungsreagenz, zur GC-Derivatisierung
Lieferant: MACHEREY-NAGEL
|
Gefahr
|
Synonyme:
TMCS, Trimethylsilylchlorid, Trimethylchlorsilan, TMSC
701280.201
701280.650
MANA701280.201EA
56.4
EUR
MANA701280.201
MANA701280.650
Chlortrimethylsilan Silylierungsreagenz, zur GC-Derivatisierung
Chlortrimethylsilan
TMCS is often used as a catalyst with other trimethylsilyl reagents. Without additives it can be used for preparing TMS derivatives of organic acids. Together with methanol it can be used for methylation.
- Silylation can be applied on many compounds
- Silylation reagents are easily prepared
- TMCS together with methanol can be also used for methylation
Formel:
C₃H₉ClSi Molecular Weight: 108,64 g/mol Siedepunkt: 57 °C (760 mm Hg) Dichte: 0,8580 g/cm³ (d20°/4°) |
MDL:
MFCD00000502 CAS-Nummer: 75-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...