N-Methyl-bis-heptafluorobutyramide (MBHFBA) Acylierungsreagenz, zur GC-Derivatisierung
Lieferant: MACHEREY-NAGEL
|
Achtung
|
701420.101
701420.201
MANA701420.101EA
379
EUR
MANA701420.101
MANA701420.201
N-Methyl-bis-heptafluorobutyramide (MBHFBA) Acylierungsreagenz, zur GC-Derivatisierung
N-Methyl-bis-heptafluorobutyramide (MBHFBA)
Acylation with fluorinated bisacylamides is recommended for alcohols, primary and secondary amines as well as for thiols under mild, neutral conditions.
- Addition of halogenated carbons enhances detectability by GC-ECD
- Derivatives are hydrolytical stable
- Increased sensitivity by adding molecular weight
Formel:
C₉H₃F₁₄NO₂ Molecular Weight: 423,10 g/mol Siedepunkt: 165…166 °C (760 mm Hg) Dichte: 1,673 g/cm³ (d20°/4°) |
MDL:
MFCD00049280 CAS-Nummer: 73980-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...