N,N-Dimethylformamiddimethylacetal Methylierungsreagenz, zur GC-Derivatisierung

Lieferant: MACHEREY-NAGEL
Gefahr

Synonyme: 1,1-Dimethoxytrimethylamin

701430.110 701430.201
MANA701430.110EA 64.7 EUR
MANA701430.110 MANA701430.201
N,N-Dimethylformamiddimethylacetal Methylierungsreagenz, zur GC-Derivatisierung
N,N-Dimethylformamiddimethylacetal

Methylation with DMF-DMA can be applied for fatty acids, primary amines and partially amino acids forming N-dimethyl-aminomethylene amino acid methyl esters. DMF-DMA is a poor solvent, for this reason it is necessary to use a mixture of DMF-DMA with pyridine, THF, acetone (barbiturates) or another solvent.


  • Methylation derivatives are generally stable
  • Wide range of reaction conditions (strongly acidic - strongly basic)
  • Some reactions can be achieved with water present

Formel: C₅H₁₃NO₂
Molecular Weight: 119,16 g/mol
Siedepunkt: 106–107 °C (760 mm Hg)
Dichte: 0,897 g/cm³ (d20°/4°)
MDL: MFCD00008482
CAS-Nummer: 4637-24-5
UN: 3316
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR