SPE columns, acrylamide analysis, CHROMABOND® ABC18
Lieferant: MACHEREY-NAGEL
730533
MANA730533EA 140 EUR
MANA730533
SPE columns, acrylamide analysis, CHROMABOND® ABC18
Chromatographiesäulen
CHROMABOND® ABC18 is a special phase for SPE analysis of acrylamide in food.
  • Special octadecyl phase
  • Additional ion exchange functions
Application: Clean-up of acrylamide from ultra-heated starch-containing food, such as potato chips, french fries, crispbread, and cereals.