Adsorbents for column chromatography, Florisil®
Lieferant: MACHEREY-NAGEL
815710.1
MANA815710.1EA
240
EUR
MANA815710.1
Adsorbents for column chromatography, Florisil®
Chromatographie-Sorbentien
HPLC-Sorbentien
Hard granular magnesia silica gel: MgO (15,5 ±0,5%) SiO2 (84,0 ±0,5%) Na2SO4 (≤1,0%) 60/100 mesh.
Typical application: sample preparation (SPE); clean-up of pesticide residues, separation of chlorinated pesticides, extraction of steroids, sex hormones, antibiotics, lipids etc.