TLC adsorbents, silica gel
Lieferant: MACHEREY-NAGEL
816320.1 816310.1 816310.5 816320.5 816330.1 816330.5 816340.1 816340.5 816380.1 816380.5 816400.1 816400.5 816410.1 816410.5
MANA816320.1EA 88.7 EUR
MANA816320.1 MANA816310.1 MANA816310.5 MANA816320.5 MANA816330.1 MANA816330.5 MANA816340.1 MANA816340.5 MANA816380.1 MANA816380.5 MANA816400.1 MANA816400.5 MANA816410.1 MANA816410.5
TLC adsorbents, silica gel
Chromatographie-Sorbentien TLC-Sorbentien
Silica adsorbents for thin layer chromatography.
  • Available in different quality grades
  • Adsorbents with or without binder
  • Available with or without fluorescence indicator
Pore volume: 0,75 ml/g
Specific surface (BET): 500 m²/g
Bestellen
TECHNISCHE DATEN
Environmentally Preferable