NANOCOLOR® Photometric Standard Tests, Manganese
Lieferant: MACHEREY-NAGEL
91860 918126
MANA91860EA 190 EUR
MANA91860 MANA918126
NANOCOLOR® Photometric Standard Tests, Manganese
Umwelt-Prüfkits
Photometrisches Testkit zur Bestimmung von Mangan in Wasserproben.
  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations
The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent: No
Digestion: No
NanOx metal: No
NanOx: No
Bestellinformation:
Lieferumfang:
Rugged box with reagents for 250 tests.