Compact photometer, PF-12Plus
Lieferant: MACHEREY-NAGEL
919250
MANA919250EA 1250 EUR
MANA919250
Compact photometer, PF-12Plus
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
Compact photometer with microprocessor control, self test and automatic calibration.
  • More than 100 preprogrammed VISOCOLOR® ECO tests and NANOCOLOR® tube tests and photometric functions
  • User-defined methods, increased flexibility with nephelometric turbidity measurements
  • Backlit graphic display with intuitive user guidance
  • Fast and easy access to stored results and data sets
  • Optimised battery indicator
Clear memory management provides storage of results with supplementary information such as date, time, sample number, sample location and dilution. The automatic filter wheel with seven interference filters, insensitive to external light for fast measurements.
Lieferumfang: Supplied with software DVD, manual, four batteries, four empty test tubes, funnel, beak, syringe, USB cable, calibration cuvette and certificate in case.