Photometric tube tests, NANOCOLOR® Organic complexing agents
Lieferant: MACHEREY-NAGEL
985052
MANA985052EA 61 EUR
MANA985052
Photometric tube tests, NANOCOLOR® Organic complexing agents
Umwelt-Prüfkits
A pre-dosed NANOCOLOR® tube test for photometric determination of complexing agents in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.