Photometric tube tests, NANOCOLOR® Organic complexing agents
Lieferant: MACHEREY-NAGEL
985052
MANA985052EA
61
EUR
MANA985052
Photometric tube tests, NANOCOLOR® Organic complexing agents
Umwelt-Prüfkits
A pre-dosed NANOCOLOR® tube test for photometric determination of complexing agents in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.