NANOCOLOR® Photometric Tube Tests, Nitrite
Lieferant: MACHEREY-NAGEL
985068 985069
MANA985068EA 74.7 EUR
MANA985068 MANA985069
NANOCOLOR® Photometric Tube Tests, Nitrite
Umwelt-Prüfkits
Photometrisches Testkit zur Bestimmung von Nitrit in Wasserproben.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent: No
Digestion: No
NanOx metal: No
NanOx: No
Bestellinformation:
Lieferumfang:
Rugged box with 20 test tubes. Sufficient for 20 tests.