NANOCOLOR® Photometric Tube Tests, Sulfite
Lieferant: MACHEREY-NAGEL
985089 985090
MANA985089EA 90.4 EUR
MANA985089 MANA985090
NANOCOLOR® Photometric Tube Tests, Sulfite
Umwelt-Prüfkits
Photometrisches Testkit zur Bestimmung von Sulfit in Wasserproben.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO easy VIS and easy UV. Programming data for other spectrophotometers are available.
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Bestellinformation:
Lieferumfang: Remark: Measuring range on NANOCOLOR VIS II