Photometric tube tests, NANOCOLOR® aluminium
Lieferant: MACHEREY-NAGEL
985098
MANA985098EA 80.5 EUR
MANA985098
Photometric tube tests, NANOCOLOR® aluminium
Umwelt-Prüfkits
A pre-dosed NANOCOLOR tube test for photometric determination of aluminium in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well."