Microscope cover glasses, 13 mm , borosilicate glass D 263 T eco, thickness No. 3 (0.28-0.32 mm) 1 * 500 ST
Lieferant: MARIENFELD
Microscope cover glasses, 13 mm , borosilicate glass D 263 T eco, thickness No. 3 (0.28-0.32 mm) 1 * 500 ST