Zählkammern, Fuchs-Rosenthal
Lieferant: MARIENFELD
0640411 0640430 0640431
MARI0640411EA 56.15 EUR
MARI0640411 MARI0640430 MARI0640431
Zählkammern, Fuchs-Rosenthal
Objektträger Zählkammern
These counting chambers with double net ruling are suitable for blood tests in vitro diagnostic (IVD) applications.
Standards: IVD directive 98/79/EC
Lieferumfang: Supplied with two cover glasses.