Zählkammern, Nageotte
Lieferant: MARIENFELD
0640511 0640530
MARI0640511EA 132.4 EUR
MARI0640511 MARI0640530
Zählkammern, Nageotte
Objektträger Zählkammern
These counting chambers are designed for leucocytes and erythrocytes in in vitro applications.
Standards: IVD directive 98/79/EC.