Microscope slide, UniMark®
Lieferant: MARIENFELD
0704022
MARI0704022EA 413.62 EUR
MARI0704022
Microscope slide, UniMark®
Objektträger Mikroskop-Objektträger
UniMark slide is designed with a white circle of Ø 12 mm and a white marking area of 20 mm for use in cytocentrifuges.
It is suitable for suitable for marking with inkjet of laser printers and permanent markers.
Standards: DIN 8037/1. IVD 98/79/EC.
Verpackung: 50 pieces in a box.