Adhesion slide, HistoBond® +S
Lieferant: MARIENFELD
0810501
MARI0810501EA 637.8 EUR
MARI0810501
Adhesion slide, HistoBond® +S
Objektträger Adhäsions-Objektträger
This adhesion slide has been designed for mounting tissue sections.
  • Autoclavable
  • Marking area
  • 90° ground edges