Adhesion slides, HistoBond®
Lieferant: MARIENFELD
0810701
MARI0810701EA 624.2 EUR
MARI0810701
Adhesion slides, HistoBond®
Objektträger Adhäsions-Objektträger
HistoBond slides combine frosted ends with printed in various colours.
  • Soda lime glass of hydrolytic class 3
  • Different colours for colour coding of methods of analysis
  • Thin-coated marking area facilitates the processing on automated systems
Standards: IVD directive 98/79/EC