Microscopy slides with 45° ground corners
Lieferant: MARIENFELD
1000812 1000912
MARI1000812EA 4.74 EUR
MARI1000812 MARI1000912
Microscopy slides with 45° ground corners
Objektträger Mikroskop-Objektträger
Microscopy slides with chamfered corners to reduce the risk of injuries and a silky area of 20mm, on one end, at both sides.
Standards: IVD 98/79/EC