Drigalski spreader
Lieferant: MARIENFELD
1800024
MARI1800024EA 33.5 EUR
MARI1800024
Drigalski spreader
Schaber Zellschaber
Glass.
  • For bacteriological applications
  • Facilitates uniform distribution and plating of bacteria material on solid nutrient media