Micro pipette controller
Lieferant: MARIENFELD
2960408
MARI2960408EA 66.35 EUR
MARI2960408
Micro pipette controller
Pipetten Pipettierhelfer
This micro pipette filler is suitable for micro pipettes with circular mark and other small pipettes up to 1 ml capacity (for example: blood diluting pipettes).
  • Facilitates the sampling
  • Easy to use
  • Integrated ejection device reduces the risk of infection through contaminated pipettes
  • Autoclavable at 121 °C