Pasteur pipettes, disposable
Lieferant: MARIENFELD
3233050 3233049
MARI3233050EA 71.3 EUR
MARI3233050 MARI3233049
Pasteur pipettes, disposable
Pipetten Pasteur- und Transfer-Pipetten
Pasteur pipettes made of soda-lime glass.
  • For single use
  • Long, fine tip
  • Constricted upper suction end to retain a cotton plug (supplied without cotton plug)
  • Handy packaging in boxes containing 250 pieces
Standards: In compliance with DIN ISO 7712