Test tubes with rim
Lieferant: MARIENFELD
3700323 3700318 3700334
MARI3700323EA 40.2 EUR
MARI3700323 MARI3700318 MARI3700334
Test tubes with rim
Röhrchen Reagenzgläser- und Probenröhrchen
Soda-Lime glass.
  • Very good chemical resistance
  • Good temperature resistance