Centrifuge tubes
Lieferant: MARIENFELD
3910002 3910012 3900022 3910022
MARI3910002EA 65.75 EUR
MARI3910002 MARI3910012 MARI3900022 MARI3910022
Centrifuge tubes
Röhrchen Zentrifugenröhrchen
Soda lime glass or borosilicate glass 3,3.
  • Graduated or ungraduated
  • With beaded or straight rim
  • Good chemical resiostance
Long conical type.