Centrifuge tubes
Lieferant: MARIENFELD
3939521
MARI3939521EA 317 EUR
MARI3939521
Centrifuge tubes
Röhrchen Zentrifugenröhrchen
Borosilicate glass 3,3.
  • Conical type
  • Blue graduated up to 20 ml, with mark at 50 ml
  • Very good chemical resistance
  • Very good temperature resistance
Centrifuge tubes for determination of solubility acc. to ADPI.