Culture media in bottles, CHROMagar™
Lieferant: MASTER AND OWNERS, R/V THOMPSON
15EC168
MAST15EC168EA
0
EUR
MAST15EC168
Culture media in bottles, CHROMagar™
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
With traditional media identification of mixed cultures is difficult, whereas CHROMagar™ just by the colour of the colony will instantly differentiate various species.
- High contrast
- Fast
- Convenient