Sealing mats, ABgene® SmartCap
Lieferant: THERMO MATRIX TECHNOLOGIES
AB-1413
MATRAB-1413EA 155 EUR
MATRAB-1413
Sealing mats, ABgene® SmartCap
Versiegeln Mikrotiterplatten-Dichtungen
Abgene™ SmartCap mat, made from advanced thermoplastic elastomer (TPE), rapidly seals 96 samples at one time in a high-density storage format, affording maximum space saving benefits. Suitable for use with Thermo Scientific™ storage tubes ranging from 0,3 ml to 1,4 ml, the Abgene™ SmartCap Mat maintains a consistent seal down to –80 °C.
  • Excellent chemical resistance and low vapour transmission rates, minimising evaporation and water uptake in DMSO-based samples
  • Samples can be accessed in full rows of eight tubes or individually with the Abgene™ SmartCap removal tool, to effortlessly remove or re-install the caps