Storage tubes, 2D barcoded
Lieferant: THERMO MATRIX TECHNOLOGIES
AB-1470
MATRAB-1470EA 904 EUR
MATRAB-1470
Storage tubes, 2D barcoded
Vials
Unique and laser etched 2D barcode is encapsulated on the base to securely identify and track samples.
  • Grid-referenced lids enable storage tube location
  • External threads tubes aid in contamination prevention
  • Save space with dense microplate footprint
The laser-etched 2D barcode feature scan the tube into any application or database, or integrate via various data export options. Every storage tube code is scanned to guarantee readability. Each code is checked against the entire database of all 2D barcoded storage tubes to ensure zero duplicates. Every lot of storage tubes is leak tested to ensure the integrity and security of samples. All tubes are certified free of DNase, RNase and DNA.