4-Methyl-2-chinolinol
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Lepidinol, 4-Methylquinolin-2-ol, 2-Hydroxy-4-methylquinoline
AC12157.1
MAYBAC12157.1EA
0
EUR
MAYBAC12157.1
4-Methyl-2-chinolinol
4-Methyl-2-chinolinol
Formel:
C₁₀H₉NO Molecular Weight: 159,185 g/mol Siedepunkt: 346 °C (1013 hPa) Schmelzpunkt: 220…222 °C |
MDL:
MFCD00006745 CAS-Nummer: 607-66-9 EINECS: 210-139-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...