5-Methylthiophen-2-carbaldehyd

Lieferant: Thermo Scientific

Synonyme: 5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde

AC12787.50
MAYBAC12787.50EA 0 EUR
MAYBAC12787.50
5-Methylthiophen-2-carbaldehyd
5-Methylthiophen-2-carbaldehyd
Formel: C₆H₆OS
Molecular Weight: 126,18 g/mol
MDL: MFCD00005434
CAS-Nummer: 13679-70-4
EINECS: 237-178-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR