5-Methyl-2-thenoesäure

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Methylthiophene-2-carboxylic acid

AC12788DA AC12788EA AC12788.50
MAYBAC12788DAEA 0 EUR
MAYBAC12788DA MAYBAC12788EA MAYBAC12788.50
5-Methyl-2-thenoesäure
5-Methyl-2-thenoesäure
Formel: C₆H₆O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00005439
CAS-Nummer: 1918-79-2
EINECS: 217-640-3

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 136°C to 139°C
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR