4-(1H-Imidazol-1-yl)phenol

Lieferant: Thermo Scientific

Synonyme: 4-(1-Imidazolyl)phenol, 4-(Imidazol-1-yl)phenol

AC17979.1
MAYBAC17979.1EA 0 EUR
MAYBAC17979.1
4-(1H-Imidazol-1-yl)phenol
4-(1H-Imidazol-1-yl)phenol
Formel: C₉H₈N₂O
Molecular Weight: 160,18 g/mol
MDL: MFCD00005281
CAS-Nummer: 10041-02-8
EINECS: 233-121-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR