3-Brom-2-chlorthiophen

Lieferant: Thermo Scientific
Achtung

AC25875DA AC25875.10
MAYBAC25875DAEA 0 EUR
MAYBAC25875DA MAYBAC25875.10
3-Brom-2-chlorthiophen
3-Bromo-2-chlorothiophene
Formel: C₄H₂BrClS
Molecular Weight: 197,48 g/mol
Siedepunkt: 195 °C (1013 hPa)
Dichte: 1,82 g/cm³ (20 °C)
MDL: MFCD00043883
CAS-Nummer: 40032-73-3
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR