3-Phenyl-1H-pyrazol
Lieferant: Thermo Scientific
Synonyme:
3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole
AC29603.50
MAYBAC29603.50EA
0
EUR
MAYBAC29603.50
3-Phenyl-1H-pyrazol
3-Phenyl-1H-pyrazole
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol |
MDL:
MFCD00159654 CAS-Nummer: 2458-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...