5-Amino-1H-indazol
Lieferant: Thermo Scientific
AC30454.10
AC30454.50
MAYBAC30454.10EA
0
EUR
MAYBAC30454.10
MAYBAC30454.50
5-Amino-1H-indazol
5-Amino-1H-indazole
Formel:
C₇H₇N₃ Siedepunkt: 345 °C (1013 hPa) Schmelzpunkt: 183 °C |
MDL:
MFCD00037975 CAS-Nummer: 19335-11-6 EINECS: 242-971-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...