2,6-Difluorphenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 1,3-Difluor-2-isothiocyanatobenzol

AC30815.10
MAYBAC30815.10EA 0 EUR
MAYBAC30815.10
2,6-Difluorphenylisothiocyanat
2,6-Difluorphenylisothiocyanat
Formel: C₇H₃F₂NS
Molecular Weight: 171,17 g/mol
Siedepunkt: 106…107 °C (35 mmHg)
Lagertemperatur: Kühlschrank
MDL: MFCD00041047
CAS-Nummer: 207974-17-2
UN: 2930
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Low melting solid or liquid
Infrared spectrum Conforms
Refractive index 1.4945 to 1.4965 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR