4-Chlor-2-(trifluormethylphenyl)isocyanat
Lieferant: Thermo Scientific
AC30912.1
AC30912.10
AC30912.50
MAYBAC30912.1EA
0
EUR
MAYBAC30912.1
MAYBAC30912.10
MAYBAC30912.50
4-Chlor-2-(trifluormethylphenyl)isocyanat
4-Chlor-2-(trifluormethylphenyl)isocyanat
Formel:
C₈H₃ClF₃NO Siedepunkt: 85…90 °C (13 mmHg) Dichte: 1,4666 g/cm³ (20 °C) |
MDL:
MFCD00013873 CAS-Nummer: 16588-69-5 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...