3,5-Bis(trifluormethyl)-1H-pyrazol

Lieferant: Thermo Scientific

Synonyme: 3,5-Bis(trifluormethyl)-1H-pyrazol

AC31181.10
MAYBAC31181.10EA 0 EUR
MAYBAC31181.10
3,5-Bis(trifluormethyl)-1H-pyrazol
3,5-Bis(trifluormethyl)-1H-pyrazol
Formel: C₅H₂F₆N₂
Molecular Weight: 204.07 g/mol
Siedepunkt: 148
MDL: MFCD00153672
CAS-Nummer: 14704-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR