2,4-Dimethoxyphenylisocyanat

Lieferant: Thermo Scientific

Synonyme: 1-Isocyanato-2,4-dimethoxybenzene

AC31429DA AC31429.50
MAYBAC31429DAEA 0 EUR
MAYBAC31429DA MAYBAC31429.50
2,4-Dimethoxyphenylisocyanat
2,4-Dimethoxyphenylisocyanat
Formel: C₉H₉NO₃
Molecular Weight: 179,18 g/mol
Siedepunkt: 140 °C (11 mmHg)
Schmelzpunkt: 30…31 °C
MDL: MFCD00002005
CAS-Nummer: 84370-87-6
EINECS: 282-747-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR